missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DR3/TNFRSF25 Antibody (JD3) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-09115H
This item is not returnable.
View return policy
Description
DR3/TNFRSF25 Monoclonal antibody specifically detects DR3/TNFRSF25 in Human samples. It is validated for Flow Cytometry
Specifications
| DR3/TNFRSF25 | |
| Monoclonal | |
| HRP | |
| Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1 | |
| human DR3/TNFRSF25-Ig fusion protein | |
| 0.1 mL | |
| Apoptosis, Cell Biology | |
| 8718 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| JD3 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction