missing translation for 'onlineSavingsMsg'
Learn More

DR3/TNFRSF25 Antibody (JD3) - BSA Free, Novus Biologicals™

Product Code. 18354012 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354012 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354012 Supplier Novus Biologicals Supplier No. NBP309115

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DR3/TNFRSF25 Monoclonal specifically detects DR3/TNFRSF25 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DR3/TNFRSF25
Applications Flow Cytometry
Classification Monoclonal
Clone JD3
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-4 ug/ml
Formulation PBS
Gene Alias Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1
Host Species Mouse
Immunogen human DR3/TNFRSF25-Ig fusion protein
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 8718
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.