missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DR3/TNFRSF25 Antibody (59204) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB943W
This item is not returnable.
View return policy
Description
DR3/TNFRSF25 Monoclonal antibody specifically detects DR3/TNFRSF25 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| DR3/TNFRSF25 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 59204 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1 | |
| Mouse myeloma cell line NS0-derived recombinant human DR3 isoform 1, Gln25-Phe201, Accession # Q93038 | |
| 0.1 mL | |
| Apoptosis, Cell Biology | |
| 8718 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction