Learn More
Invitrogen™ DR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589713
Description
Immunogen sequence: QGGTRSPRCD CAGDFHKKIG LFCCRGCPAG HYLKAPCTEP CGNSTCLVCP QDTFLAWENH HNSECARCQA CDEQASQVAL ENCSAVADTR CGCKPGWFVE CQVSQCVSSS PFYCQPCLDC GALHRHTRLL CSRRDTDCGT CLPGFYEHGD GCVSCPTPPP SLAGAP Positive Samples: Raji, HT-29, U-87MG, A-549; Cellular Location: Cell membrane, Single-pass type I membrane protein.
DR3/TNFRSF25 is a member of the TNF-receptor superfamily. This receptor is expressed preferentially in the tissues enriched in lymphocytes, and it may play a role in regulating lymphocyte homeostasis. This receptor has been shown to stimulate NF-kappa B activity and regulate cell apoptosis. The signal transduction of this receptor is mediated by various death domain containing adaptor proteins. Knockout studies in mice suggested the role of DR3/TNFRSF25 in the removal of self-reactive T cells in the thymus. Multiple alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported, most of which are potentially secreted molecules. The alternative splicing of this gene in B and T cells encounters a programmed change upon T-cell activation, which predominantly produces full-length, membrane bound isoforms, and is thought to be involved in controlling lymphocyte proliferation induced by T-cell activation.
Specifications
| DR3 | |
| Polyclonal | |
| Unconjugated | |
| TNFRSF25 | |
| APO3; APO-3; apoptosis inducing receptor; apoptosis-inducing receptor AIR; Apoptosis-mediating receptor DR3; apoptosis-mediating receptor TRAMP; DDR3; death domain receptor 3 soluble form; Death receptor 3; death receptor beta; DR3; LARD; lymphocyte-associated receptor of death; Protein WSL; Protein WSL-1; RP4-650H14.2; TNF receptor superfamily member 25; TNFRSF12; TNFRSF25; TR3; TRAMP; tumor necrosis factor receptor superfamily member 25; tumor necrosis factor receptor superfamily, member 12; tumor necrosis factor receptor superfamily, member 12 (translocating chain-association membrane protein); tumor necrosis factor receptor superfamily, member 25; UNQ455/PRO779; Wsl; WSL1; WSL-1; WSL-LR | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 500592, 85030, 8718 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.69 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q93038 | |
| TNFRSF25 | |
| Recombinant Protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.