missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109659
This item is not returnable.
View return policy
Description
Immunogen sequence: MASSSGNDDD LTIPRAAINK MIKETLPNVR VANDARELVV NCCTEFIHLI SSEANEICNK SEKKTISPEH VIQALESLGF GSYISEVKEV LQECKTVALK RRKASSRLEN LGIPEEELLR QQQELFAKAR QQQAELAQQE WLQMQQAAQQ AQLAAASASA SNQAGSSQDE EDDDDI.
This gene encodes a TBP- (TATA box-binding protein) associated phosphoprotein that represses both basal and activated levels of transcription. The encoded protein is phosphorylated in vivo and this phosphorylation affects its interaction with TBP. This protein contains a histone fold motif at the amino terminus, a TBP-binding domain, and a glutamine- and alanine-rich region. The binding of DR1 repressor complexes to TBP-promoter complexes may establish a mechanism in which an altered DNA conformation, together with the formation of higher order complexes, inhibits the assembly of the preinitiation complex and controls the rate of RNA polymerase II transcription.
Specifications
| DR1 | |
| Polyclonal | |
| Unconjugated | |
| Dr1 | |
| 1700121L09Rik; Down-regulator of transcription 1; down-regulator of transcription 1, TBP-binding (negative cofactor 2); Dr1; Dr1l; NC2; NC2B; NC2beta; NC2-beta; negative cofactor 2; negative cofactor 2 beta; negative cofactor 2-beta; Protein Dr1; similar to down-regulator of transcription 1; TATA-binding protein-associated phosphoprotein; wu:fj15f04; zgc:152914 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13486, 1810, 289881 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.88 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q01658, Q5XI68, Q91WV0 | |
| Dr1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-176 of human DR1 (NP_001929.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction