missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DR1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97557AF350
This item is not returnable.
View return policy
Description
DR1 Polyclonal specifically detects DR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| DR1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| Down-regulator of transcription 1, down-regulator of transcription 1, TBP-binding (negative cofactor 2), NC2, NC2-BETA, Negative cofactor 2-beta, protein Dr1, TATA-binding protein-associated phosphoprotein | |
| Produced in rabbits immunized with E. coli-derived Human DR1 fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 1810 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction