missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ X-plore™ 3300 Half-Mask Respirators

Product Code. p-3838158
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10532783

Brand: Dräger™ R55331

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Made from soft-TPE material, X-plore™ half-mask respirators offer an innovative, modern design to ensure easy, comfortable breathing.

  • Modern, low-profile mask body design
  • Extended field of vision and room to easily use visors and shields
  • Can be worn comfortably under helmets and visors
  • Compact, lightweight and consists of fewer parts
  • Soft, lightweight and flexible strap system
  • Flexi-Fit head harness conforms to the contours of head without pressure or hair entanglement
  • Innovative X-Guided cross-over strap system ensure even weight distribution and an even seal
  • 2-point bayonet connector with markings for insertion of the filter
  • Flexible nose seal fits securely with safety glasses and goggles
  • Insert molding technology, smooth surface area
  • Easy to disassemble, clean and reassemble

Specifications

Certifications/Compliance AS NZS 1716
Certifications (EN) EN 140
Certifications (NIOSH) NIOSH 42 CFR Part 84
Connection Type Bayonet
Color Blue
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Small
For Use With (Equipment) X-plore™ Bayonet Filters and Cartridges
Model R55331
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.