Learn More
Dräger™ Short-Term Detector Tubes - Phosgene
Product Code. p-4909573
Change view
Click to view available options
Components Measured:
Phosgene 0.02/a
Phosgene 0.25/b
Measurement Range:
0.02 to 1ppm
0.25 to 5ppm
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Measurement Range unitSize
13197276 Phosgene 0.25/b 0.25 to 5ppm Pack of 10
11368291 Phosgene 0.02/a 0.02 to 1ppm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13197276 Supplier Dräger™ Supplier No. CH28301
  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.
Specifications
Components Measured Phosgene 0.25/b
Humidity Range 3 to 15mg/L
Measurement Range 0.25 to 5ppm
No. of Strokes 5
Quantity 10
Response Time 1 min.
Temperature Range (English) 41°F to 95°F
Temperature Range (Metric) 5° to 35°C