missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes - Nitric Acid

Product Code. 10174863
Change view
Click to view available options
Measurement Range:
1 to 50ppm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range unitSize
10174863 1 to 50ppm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10174863 Supplier Dräger™ Supplier No. 6728311

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.

Specifications

Components Measured Nitric Acid 1/a
Humidity Range 3 to 15mg/L
Measurement Range 1 to 50ppm
No. of Strokes 10/20
Quantity 10
Response Time 2 to 4 min.
Temperature Range (English) 41°F to 104°F
Temperature Range (Metric) 5° to 40°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.