missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes - De Tube Monostyrene

Product Code. 12334579
Click to view available options
Components Measured:
Monostyrene 10/a
Monostyrene 10/b
Measurement Range:
10 to 200ppm
10 to 250ppm
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12334579

Brand: Dräger™ 6733141

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.

Specifications

Components Measured Monostyrene 10/b
Humidity Range 3 to 15mg/L
Measurement Range 10 to 250ppm
No. of Strokes 20
Quantity 10
Response Time 3 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
compliance-icons compliance-icons
Product Identifier
  • Dräger-Tubes
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.