Learn More
Dräger™ Short-Term Detector Tubes - De Tube Mercaptan
Brand: Dräger™ 6728981
Code : 50
Additional Details : Weight : 0.07740kg
Description
1Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
2Safety Equipment Institute (SEI) certified.
Specifications
Mercaptan 0.5/a | |
0.5 to 5ppm | |
10 | |
50°F to 104°F |
3 to 15mg/L | |
20 | |
5 min. | |
10° to 40°C |
Safety and Handling
- X10 Tube Mercaptan 0.5a range 0.5-5ppm 0.5 to 5 ppm0.5 to 5 ppm+/- 10 to 15% (pack of 10)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Supplemental information
- MIXTURE LIST-Contains : mercaptan