Learn More
Dräger™ Short-Term Detector Tubes: Formaldehyde
Detector tubes
£105.00 - £210.53
Specifications
Components Measured | Formaldehyde |
---|---|
Humidity Range | 3 to 15mg/L |
Quantity | 10 |
Product Code | Brand | Components Measured | Tube Type | Measurement Range | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Components Measured | Tube Type | Measurement Range | Price | Quantity & Availability | |||
10059050
|
Dräger™
6733081 |
Formaldehyde | Formaldehyde 0.2/a | 0.2 to 5 |
£210.53
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12712517
|
Dräger™
8101751 |
Formaldehyde | Formaldehyde 2/a | 2 to 40 |
£105.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Formaldehyde | |
3 to 15mg/L | |
10 |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H332-Harmful if inhaled.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.