Learn More
Dräger™ Short-Term Detector Tubes: Dimethyl Sulfide
Artikelnummer. 12818270
Skift visning
Klik for at se tilgængelige muligheder
Measurement Range:
1 to 15
Pakningsstørrelse:
Each
Artikelnummer. Measurement Range unitSize
12818270 1 to 15 Each
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 12818270 Leverandør Dräger™ Leverandørnr. 6728451
Denne vare kan ikke returneres. Se returpolicy
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Ordreinfo
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Components Measured Dimethyl Sulfide
Humidity Range Less than20mg/L
Measurement Range 1 to 15
No. of Strokes 20
Quantity 5
Response Time 15 min.
Temperature Range (English) 59°F to 86°F
Temperature Range (Metric) 15°C to 30°C
Tube Type Dimethyl sulfide 1/a