Learn More
Dräger™ Short-Term Detector Tubes: Dimethyl Formamide
Product Code. 10764635
Change view
Click to view available options
Measurement Range:
10 to 40
Unit Size:
Pack of 10
Product Code. Measurement Range unitSize
10764635 10 to 40 Pack of 10
1 options
Product Code. 10764635 Supplier Dräger™ Supplier No. 6718501
  • ONLINE EXCLUSIVE PRICE
    £200.55
    £170.73 / Pack of 10
    Save £29.82
    14% Off
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Components Measured Dimethyl Formamide
Humidity Range 3 to 12mg/L
Measurement Range 10 to 40
No. of Strokes 10
Quantity 10
Response Time 3 min.
Temperature Range (English) 59°F to 95°F
Temperature Range (Metric) 15° to 35°C
Tube Type Dimethyl formamide 10/b