missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes: Sulfur Dioxide

Product Code. 10632693
Change view
Click to view available options
Measurement Range:
0.1 to 3
0.5 to 25
1 to 25
20 to 2000
50 to 8000
Tube Type:
Sulfur dioxide 0.1/a
Sulfur dioxide 0.5/a2
Sulfur dioxide 1/a
Sulfur dioxide 20/a
Sulfur dioxide 50/b
Response Time:
20 min.
3 min.
3 to 6 min.
Not Applicable
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Tube Type Response Time Unit Size
10632693 0.1 to 3 Sulfur dioxide 0.1/a 20 min. Pack of 10
10482452 50 to 8000 Sulfur dioxide 50/b Not Applicable Pack of 10
10153312 20 to 2000 Sulfur dioxide 20/a 3 min. Pack of 10
10127390 0.5 to 25 Sulfur dioxide 0.5/a2 3 to 6 min. Pack of 10
10206731 1 to 25 Sulfur dioxide 1/a 3 min. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10632693 Supplier Dräger™ Supplier No. 6727101
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detector tubes

Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.

Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.

Order Info

Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.

Specifications

Components Measured Sulfur Dioxide
Humidity Range 3 to 15mg/L
Measurement Range 0.1 to 3
No. of Strokes 100
Quantity 10
Response Time 20 min.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C
Tube Type Sulfur dioxide 0.1/a
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.