missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Pac™ III Sensors

Product Code. p-3888245
Click to view available options
Components Measured:
Hydrogen Sulfide (XS)
Oxygen (XS)
Measurement Range:
0 to 100
0 to 25%
Warranty:
3 Years
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11362080

Brand: Dräger™ 6809130

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provide fast and accurate responses to hazardous gas levels

  • For use with Pac™ III single gas monitors

Specifications

Components Measured Oxygen (XS)
Humidity Range 10 to 95%
Measurement Range 0 to 25%
Pressure Range 700 to 1300mbar
Resolution 0.1%
Temperature Range (English) -40°F to +122°F
Temperature Range (Metric) -40° to +50°C
Warranty 3 Years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.