missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DQX1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15537671
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15537671 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15537671 Supplier Invitrogen™ Supplier No. PA536432

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 79 kDa. Purity is >95% by SDS-PAGE.

DQX1 (DEAQ box RNA-dependent ATPase 1), also known as FLJ23757, is a 71 amino acid protein that contains one helicase ATP-binding domain and one helicase C-terminal domain. Localized to the nucleus, DQX1 catalyzes the conversion of ATP to ADP and a phosphate. Expressed as three isoforms produced by alternative splicing events, DQX1 is encoded by a gene that maps to human chomosome 2. As the second largest human chromosome, chromosome 2 makes up approximately 8% of the human genome and contains 237 million bases encoding over 1,400 genes. A number of genetic diseases are linked to genes on chromosome 2, including Harlequin icthyosis, sitosterolemia and Alstrom syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DQX1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene DQX1
Gene Accession No. Q8TE96, Q924H9
Gene Alias 2310066E11Rik; ATP-dependent RNA helicase DQX1; DEAQ box polypeptide 1; DEAQ box polypeptide 1 (RNA-dependent ATPase); DEAQ box RNA-dependent ATPase 1; DEAQ RNA-dependent ATPase; DEAQ RNA-dependent ATPase DQX1; DEAQ-box RNA dependent ATPase 1; Dqx1
Gene Symbols DQX1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 560-610 of human DQX1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165545, 680434, 93838
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.