missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DQX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | DQX1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
DQX1 Polyclonal specifically detects DQX1 in Human samples. It is validated for Western Blot.Specifications
| DQX1 | |
| Polyclonal | |
| Rabbit | |
| ATP-dependent RNA helicase DQX1, DEAQ box polypeptide 1, DEAQ box polypeptide 1 (RNA-dependent ATPase), DEAQ box RNA-dependent ATPase 1, DEAQ RNA-dependent ATPase DQX1, EC 3.6.4.12, FLJ23757 | |
| DQX1 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 165545 | |
| Synthetic peptides corresponding to DQX1(DEAQ box polypeptide 1 (RNA-dependent ATPase)) The peptide sequence was selected from the N terminal of DQX1. Peptide sequence LRFCWDRLLLQEVASTRGTGAWGVLVLDEAQERSVASDSLQGLLQDARLE. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title