missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPYSL4 Rabbit anti-Human, Rat, Clone: 8N2S10, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | DPYSL4 |
|---|---|
| Clone | 8N2S10 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18387721
|
Novus Biologicals
NBP3-15288-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18381355
|
Novus Biologicals
NBP3-15288-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DPYSL4 Monoclonal antibody specifically detects DPYSL4 in Human, Rat samples. It is validated for Western BlotSpecifications
| DPYSL4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 10570 | |
| IgG | |
| Affinity purified |
| 8N2S10 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| CRMP-3, CRMP3Collapsin response mediator protein 3, dihydropyrimidinase-like 4, dihydropyrimidinase-related protein 4, DRP-4UNC33-like phosphoprotein 4, ULIP4ULIP-4 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human DPYSL4 (O14531). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto