missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPYS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576917
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Dihydropyrimidinase catalyzes the conversion of 5,6-dihydrouracil to 3-ureidopropionate in pyrimidine metabolism. Dihydropyrimidinase is expressed at a high level in liver and kidney as a major 2.5-kb transcript and a minor 3.8-kb transcript. Defects in the DPYS gene are linked to dihydropyrimidinuria.
Specifications
| DPYS | |
| Polyclonal | |
| Unconjugated | |
| Dpys | |
| 1200017I10Rik; 1300004I01Rik; DHP; DHPase; dihydropyrimidinase; Dihydropyrimidine amidohydrolase; Dpys; Hydantoinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1807, 64705, 65135 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14117, Q63150, Q9EQF5 | |
| Dpys | |
| Recombinant full length Human DPYS. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction