missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPYS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13237-1-AP-150UL
This item is not returnable.
View return policy
Description
Dihydropyrimidinase catalyzes the conversion of 5,6-dihydrouracil to 3-ureidopropionate in pyrimidine metabolism. Dihydropyrimidinase is expressed at a high level in liver and kidney as a major 2.5-kb transcript and a minor 3.8-kb transcript. Defects in the DPYS gene are linked to dihydropyrimidinuria.Specifications
| DPYS | |
| Polyclonal | |
| Unconjugated | |
| DPYS | |
| DHP, DHPase, Dihydropyrimidinase, DPYS, Hydantoinase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1807, 64705, 65135 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14117, Q63150, Q9EQF5 | |
| DPYS | |
| DPYS Fusion Protein Ag3972 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction