missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPYS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112406
This item is not returnable.
View return policy
Description
DPYS Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Dihydropyrimidinase catalyzes the conversion of 5,6-dihydrouracil to 3-ureidopropionate in pyrimidine metabolism. Dihydropyrimidinase is expressed at a high level in liver and kidney as a major 2.5-kb transcript and a minor 3.8-kb transcript. Defects in the DPYS gene are linked to dihydropyrimidinuria.
Specifications
| DPYS | |
| Polyclonal | |
| Unconjugated | |
| Dpys | |
| 1200017I10Rik; 1300004I01Rik; DHP; DHPase; dihydropyrimidinase; Dihydropyrimidine amidohydrolase; Dpys; Hydantoinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1807, 64705 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q14117, Q9EQF5 | |
| Dpys | |
| Recombinant Human Dihydropyrimidinase protein (250-519AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction