missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPYD Recombinant Rabbit Monoclonal Antibody (23GB5195), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552815
This item is not returnable.
View return policy
Description
Involved in pyrimidine base degradation. Catalyzes the reduction of uracil and thymine. Also involved the degradation of the chemotherapeutic drug 5-fluorouracil.
Specifications
| DPYD | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DPYD | |
| AI315208; DHP; DHPDHase; dihydropyrimidine dehydrogenase; dihydropyrimidine dehydrogenase [NADP(+)]; Dihydrothymine dehydrogenase; dihydrouracil dehydrogenase; DPD; Dpyd; E330028L06Rik | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1806, 81656 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5195 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O89000, Q12882 | |
| DPYD | |
| A synthesized peptide derived from human DPYD (851-910AA). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction