missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DPYD Recombinant Rabbit Monoclonal Antibody (23GB5195), Invitrogen™
GREENER_CHOICE

Product Code. 30087406
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30087406 100 μL 100µL
30088745 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30087406 Supplier Invitrogen™ Supplier No. MA552814

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Involved in pyrimidine base degradation. Catalyzes the reduction of uracil and thymine. Also involved the degradation of the chemotherapeutic drug 5-fluorouracil.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPYD
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5195
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DPYD
Gene Accession No. O89000, Q12882
Gene Alias AI315208; DHP; DHPDHase; dihydropyrimidine dehydrogenase; dihydropyrimidine dehydrogenase [NADP(+)]; Dihydrothymine dehydrogenase; dihydrouracil dehydrogenase; DPD; Dpyd; E330028L06Rik
Gene Symbols DPYD
Host Species Rabbit
Immunogen A synthesized peptide derived from human DPYD (851-910AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1806, 81656
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.