Learn More
DPY30 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16281-1-AP-20UL
This item is not returnable.
View return policy
Description
The DPY30 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human DPY30. This antibody recognizes human, mouse, rat antigen. The DPY30 antibody has been validated for the following applications: IF, IHC,ELISA analysis.
Specifications
| DPY30 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 84661 | |
| Liquid |
| Immunohistochemistry, Immunofluorescence, ELISA | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |