missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPY30 Monoclonal Antibody (1-6)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA532900
This item is not returnable.
View return policy
Description
DPY30 Monoclonal Antibody for Western Blot, ICC/IF
As part of the MLL1/MLL complex, involved in the methylation of histone H3 at 'Lys-4', particularly trimethylation. Histone H3 'Lys-4' methylation represents a specific tag for epigenetic transcriptional activation. May play some role in histone H3 acetylation. In a teratocarcinoma cell, plays a crucial role in retinoic acid-induced differentiation along the neural lineage, regulating gene induction and H3 'Lys-4' methylation at key developmental loci. May also play an indirect or direct role in endosomal transport.
Specifications
| DPY30 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q99LT0, Q9C005 | |
| DPY30 | |
| Recombinant protein within mouse DPY30 aa 1-99. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| 1-6 | |
| Unconjugated | |
| DPY30 | |
| 2810410M20Rik; Aip1; C87842; Cps25; dosage compensation-related protein DPY30; DPY30; dpy-30 histone methyltransferase complex regulatory subunit; dpy-30 homolog; dpy-30 homolog (C. elegans); dpy-30, histone methyltransferase complex regulatory subunit; Dpy-30L; dpy-30-like protein; HDPY-30; Protein dpy-30 homolog; protein rAIP1; Saf19; Unknown (protein for MGC:136993) | |
| Mouse | |
| Protein A | |
| RUO | |
| 66310, 84661 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction