missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DPTN

Product Code. 18739426
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18739426

Brand: Tocris Bioscience™ 7628/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective A3 adenosine receptor antagonist

DPTN is a potent and selective A3 adenosine receptor antagonist (Ki values are in the range 0.36 - 1.65 nM (hA3AR) and 1.6 - 8.53 nM (rA3AR)). Reduces intrapulmonary pressure when combined with Dexamethasone (Cat. No. 1126).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[4-(3,5-Dimethylphenyl)-5-(4-pyridinyl)-2-thiazolyl]-3-pyridinecarboxamide dihydrochloride
CAS 325767-87-1
Quantity 50 mg
Target Adenosine A3 R
Molecular Formula C22H18N4OS.2HCl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.