missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPPIV/CD26 Antibody (236.3), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
£292.00
Specifications
| Antigen | DPPIV/CD26 |
|---|---|
| Clone | 236.3 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | mFluor Violet 500 SE |
Description
DPPIV/CD26 Monoclonal antibody specifically detects DPPIV/CD26 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| DPPIV/CD26 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation | |
| mFluor Violet 500 SE | |
| Mouse | |
| Adaptive Immunity, Cellular Markers, GPCR, Immunology | |
| 50 mM Sodium Borate | |
| 1803 | |
| IgG2b κ | |
| Protein G purified |
| 236.3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5 | |
| Rat 110-120 kD single chain glycoprotein Depeptidyl Peptidase IV (CD26). | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title