Learn More
DPPII/QPP/DPP7 Antibody (035), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90774PCP
This item is not returnable.
View return policy
Description
DPPII/QPP/DPP7 Monoclonal antibody specifically detects DPPII/QPP/DPP7 in Mouse samples. It is validated for ELISA
Specifications
| Monoclonal | |
| ELISA | |
| Protein A purified | |
| 29952 | |
| Purified |
| PerCP | |
| PBS | |
| Primary | |
| Store at 4C in the dark. |