missing translation for 'onlineSavingsMsg'
Learn More

DPPII/QPP/DPP7 Antibody (003), DyLight 755, Novus Biologicals™

Product Code. 18404477 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404477 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404477 Supplier Novus Biologicals Supplier No. NBP289622IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DPPII/QPP/DPP7 Monoclonal antibody specifically detects DPPII/QPP/DPP7 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPPII/QPP/DPP7
Applications ELISA
Classification Monoclonal
Clone 003
Conjugate DyLight 755
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias carboxytripeptidase, Dipeptidyl aminopeptidase II, dipeptidyl arylamidase II, dipeptidyl peptidase 2, Dipeptidyl peptidase 7, Dipeptidyl peptidase II, dipeptidylpeptidase 7, dipeptidyl-peptidase 7, dipeptidyl-peptidase II, DPP II, DPP2, DPPII, EC 3.4.14.2, QPP, Quiescent cell proline dipeptidase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human DPPII/QPP/DPP7 (NP_037511.2; Met1-Leu492).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29952
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.