missing translation for 'onlineSavingsMsg'
Learn More

DPPA2 Antibody (OTI1G10), DyLight 594, Novus Biologicals™

Product Code. 18432927 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432927 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432927 Supplier Novus Biologicals Supplier No. NBP272426DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DPPA2 Monoclonal antibody specifically detects DPPA2 in Human, Rat, Canine samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPPA2
Applications Western Blot
Classification Monoclonal
Clone OTI1G10
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias CT100, developmental pluripotency associated 2, developmental pluripotency-associated protein 2, PESCRG1cancer/testis antigen 100, Pluripotent embryonic stem cell-related gene 1 protein
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-175 of human DPPA2 (NP_620170) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Developmental Biology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 151871
Target Species Human, Rat, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.