missing translation for 'onlineSavingsMsg'
Learn More

DPPA2 Antibody [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30738162 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30738162 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30738162 Supplier Novus Biologicals Supplier No. AF7086JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

DPPA2 Polyclonal antibody specifically detects DPPA2 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPPA2
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias CT100, developmental pluripotency associated 2, developmental pluripotency-associated protein 2, PESCRG1cancer/testis antigen 100, Pluripotent embryonic stem cell-related gene 1 protein
Host Species Sheep
Immunogen E. coli-derived recombinant human DPPA2, Ser2-Thr174, Accession # Q7Z7J5
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Developmental Biology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 151871
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.