missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPP9 Antibody (OTI2E3), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72318AF594
This item is not returnable.
View return policy
Description
DPP9 Monoclonal specifically detects DPP9 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| DPP9 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| DPP9 | |
| Full length recombinant protein of human DPP9 (NP_631898) produced in HEK293T cells. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI2E3 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| dipeptidyl peptidase 9, Dipeptidyl peptidase IV-related protein 2, dipeptidyl peptidase IV-related protein-2, Dipeptidyl peptidase IX, Dipeptidyl peptidase-like protein 9, dipeptidylpeptidase 9, dipeptidyl-peptidase 9, DKFZp762F117, DP9, DPLP9, DPP IX, DPRP2, DPRP-2, EC 3.4.14.5, FLJ16073 | |
| Mouse | |
| 0.1 mL | |
| 91039 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction