missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPP8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38387-20ul
This item is not returnable.
View return policy
Description
DPP8 Polyclonal antibody specifically detects DPP8 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| DPP8 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| dipeptidyl peptidase 8, Dipeptidyl peptidase IV-related protein 1, dipeptidyl peptidase IV-related protein-1, dipeptidylpeptidase 8, dipeptidyl-peptidase 8, DP8Dipeptidyl peptidase VIII, DPP VIII, DPRP-1, DPRP1Prolyl dipeptidase DPP8, EC 3.4.14.5, FLJ14920, FLJ20283, MGC26191, MSTP141 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 10-230 of human DPP8 (NP_932065.1).,, Sequence:, IKSGKCNMAAAMETEQLGVEIFETADCEENIESQDRPKLEPFYVERYSWSQLKKLLADTRKYHGYMMAKAPHDFMFVKRNDPDGPHSDRIYYLAMSGENRENTLFYSEIPKTINRAAVLMLSWKPLLDLFQATLDYGMYSREEELLRERKRIGTVGIASYDYHQGSGTFLFQAGSGIYHVKDGGPQGFTQQPLRPNLVETSCPNIRMDPKLCPADPDWIAF | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54878 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction