missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPP6 Antibody (274308), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2360D
This item is not returnable.
View return policy
Description
DPP6 Monoclonal antibody specifically detects DPP6 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOF
Specifications
| DPP6 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1804 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunoprecipitation, CyTOF | |
| 274308 | |
| Flow Cytometry, Immunoprecipitation, CyTOF-ready | |
| dipeptidyl aminopeptidase IV-related protein, dipeptidyl aminopeptidase-like protein 6, Dipeptidyl aminopeptidase-related protein, Dipeptidyl peptidase 6, Dipeptidyl peptidase IV-like protein, dipeptidyl peptidase IV-related protein, Dipeptidyl peptidase VI, dipeptidylpeptidase 6, dipeptidyl-peptidase 6, dipeptidylpeptidase VI, DPP VI, DPPXFLJ55680, MGC46605, VF2 | |
| Mouse myeloma cell line NS0-derived recombinant human DPP6, Thr56-Asp803 (Glu458Val), Accession # AAA35761 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction