missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DPP4 Recombinant Rabbit Monoclonal Antibody (009), PE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA560480

 View more versions of this product

Product Code. 30535532

  • £82.40 / Pack of 25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DPP4 Recombinant Monoclonal Antibody for Flow

CD26 (dipeptidyl peptidase IV, DPP IV), adenosine deaminase (ADA) binding protein) is a homodimeric atypical serine protease belonging to the prolyl oligopeptidase family. CD26 is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. CD26 identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. Further, CD26 is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. Alterations in CD26 peptidase activity are characteristic of malignant transformation, and the enzymatic activity increases dramatically with tumor grade and severity. CD26 is expressed in various blood cell types, but also in cells that are histogenetically related to activated fibroblasts. Alterations in CD26 density have been reported on circulating monocytes and CD4+ T cells during rheumatoid arthritis and systemic lupus erythematosus.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DPP4
Recombinant Monoclonal
0.1 mg/mL
PBS with 0.5% BSA and 0.09% sodium azide
P28843
DPP4
Recombinant Mouse DPP4/CD26 protein (Ser29-His760).
25 Tests
Primary
Mouse
Antibody
IgG
Flow Cytometry
009
PE
DPP4
ACT3; Activation molecule 3; ADABP; ADCP2; ADCP-2; ADCP-I; adenosine deaminase complexing protein; Adenosine deaminase complexing protein 2; bile canaliculus domain-specific membrane glycoprotein; CD26; Dipeptidyl peptidase 4; Dipeptidyl peptidase 4 60 kDa soluble form; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV 60 kDa soluble form; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidylpeptidase 4; dipeptidyl-peptidase 4; dipeptidyl-peptidase 4 (CD26, adenosine deaminase complexing protein 2); dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP4; Dpp-4; DPPIV; DPP-IV; GP110 glycoprotein; I79_016618; serine protease; T-cell activation antigen CD26; THAM; Thymocyte-activating molecule; TP103; WC10
Rabbit
Protein A
RUO
13482
4°C, store in dark, DO NOT FREEZE!
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.