missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DPP4 Recombinant Rabbit Monoclonal Antibody (007), APC, Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£93.50 - £113.00

Specifications

Antigen DPP4
Clone 007
Concentration 100 μg/mL
Applications Flow Cytometry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30019875
GREENER_CHOICE
View Documents
Invitrogen™
MA552417
100 Tests
£113.00
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30019546
View Documents
Invitrogen™
MA552418
25 Tests
£93.50
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DPP4 Recombinant Monoclonal antibody specifically detects DPP4 in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

DPP4
100 μg/mL
Recombinant Monoclonal
Liquid
RUO
PBS with 0.5% BSA and 0.09% sodium azide
ACT3; Activation molecule 3; ADABP; ADCP2; ADCP-2; ADCP-I; adenosine deaminase complexing protein; Adenosine deaminase complexing protein 2; bile canaliculus domain-specific membrane glycoprotein; CD26; Dipeptidyl peptidase 4; Dipeptidyl peptidase 4 60 kDa soluble form; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV 60 kDa soluble form; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidylpeptidase 4; dipeptidyl-peptidase 4; dipeptidyl-peptidase 4 (CD26, adenosine deaminase complexing protein 2); dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP4; Dpp-4; DPPIV; DPP-IV; GP110 glycoprotein; I79_016618; serine protease; T-cell activation antigen CD26; THAM; Thymocyte-activating molecule; TP103; WC10
Recombinant Mouse DPP4/CD26 Protein (Ser29-His760).
Primary
4°C, store in dark, DO NOT FREEZE!
DPP4
007
Flow Cytometry
APC
Rabbit
Mouse
P28843
13482
IgG
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.