missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPP3 Monoclonal Antibody (OTI2D9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525629
This item is not returnable.
View return policy
Description
DPP3 Monoclonal Antibody for Western Blot, IHC (P), Flow
This gene encodes a protein that is a member of the M49 family of metallopeptidases. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH) and has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Increased activity of this protein is associated with endometrial and ovarian cancers. Alternate transcriptional splice variants have been characterized.
Specifications
| DPP3 | |
| Monoclonal | |
| 0.58 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NY33 | |
| DPP3 | |
| Full length human recombinant protein of DPP3 produced in HEK293 cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D9 | |
| Unconjugated | |
| DPP3 | |
| 4930533O14Rik; C86324; Dipeptidyl aminopeptidase III; dipeptidyl arylamidase III; Dipeptidyl peptidase 3; dipeptidyl peptidase III; dipeptidylpeptidase 3; dipeptidyl-peptidase 3; dipeptidylpeptidase III; dipeptidyl-peptidase III; DPP III; Dpp3; DPPIII; Enkephalinase B; hCG_2016942 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10072 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction