missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPP10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109655
This item is not returnable.
View return policy
Description
Immunogen sequence: GYIASMILKS DEKLFKCGSV VAPITDLKLY ASAFSERYLG MPSKEESTYQ AASVLHNVHG LKEENILIIH GTADTKVHFQ HSAELIKHLI KAGVNYTMQV Y.
A novel gene, DPP10, encodes a homolog of dipeptidyl peptidases (DPPs) that cleave terminal dipeptides from cytokines and chemokines, and it presents a potential new target for asthma therapy. The DPP10 protein shares features with members of the S9B family of DPP serine proteases, which includes DPP4, a widely expressed enzyme that plays a central role in chemokine processing as part of the innate immune system. The locus displays a complex pattern of transcript splicing, with eight alternate first axons; four of which localize within the LD block strongly associated with asthma.
Specifications
| DPP10 | |
| Polyclonal | |
| Unconjugated | |
| DPP10 | |
| 6430601K09Rik; dipeptidyl peptidase 10; dipeptidyl peptidase IV-related protein 3; dipeptidyl peptidase like 10; dipeptidyl peptidase X; Dipeptidyl peptidase-like protein 2; dipeptidylpeptidase 10; dipeptidyl-peptidase 10 (inactive); dipeptidyl-peptidase 10 (non-functional); DPL2; DPP X; DPP10; DPPY; Dprp3; DPRP-3; HGNC:20823; inactive dipeptidyl peptidase 10; KIAA1492; Kv4 potassium channel auxiliary subunit; OTTHUMP00000203618 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57628 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.52 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8N608 | |
| DPP10 | |
| A synthetic peptide corresponding to a sequence within amino acids 400-500 of human DPP10 (NP_001308842.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction