missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DPM1 Polyclonal antibody specifically detects DPM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | DPM1 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | CDGIEEC 2.4.1.83, dolichol monophosphate mannose synthase, dolichol-phosphate mannosyltransferase, Dolichyl-phosphate beta-D-mannosyltransferase, dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit, DPM synthase, Mannose-P-dolichol synthase, MPD synthase, MPDSDolichol-phosphate mannose synthase |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human DPM1 (NP_003850.1). MASLEVSRSPRRSRRELEVRSPRQNKYSVLLPTYNERENLPLIVWLLVKSFSESGINYEIIIIDDGSPDGTRDVAEQLEKIYGSDRILLRPREKKLGLGTAYIHGMKHATGNYIIIMDADLSHHPKFIPEFIRKQKEGNFDIVSGTRYKGNGGVYGWDLKRKIISRGANFLTQILLRPGASDLTGSFRLYRKEVLEKLIEKCVSKGYVFQMEMIVRARQLNYTIGEVPISFVDRVYGESKLGGNEIVSFLKGLLTLFATT |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?