missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPH5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584994
This item is not returnable.
View return policy
Description
Immunogen sequence: VLRATKLGIP YRVIHNASIM NAVGCCGLQL YKFGETVSIV FWTDTWRPES FFDKVKKNRQ NGMHTLCLLD IKVKEQSLEN LIKGRKIYEP PRYMS Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 94%.
This gene encodes a component of the diphthamide synthesis pathway. Diphthamide is a post-translationally modified histidine residue found only on translation elongation factor 2. It is conserved from archaebacteria to humans, and is targeted by diphtheria toxin and Pseudomonas exotoxin A to halt cellular protein synthesis. The yeast and Chinese hamster homologs of this protein catalyze the trimethylation of the histidine residue on elongation factor 2, resulting in a diphthine moiety that is subsequently amidated to yield diphthamide. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2008].
Specifications
| DPH5 | |
| Polyclonal | |
| Unconjugated | |
| DPH5 | |
| AD-018; CGI-30; diphthamide biosynthesis 5; diphthamide biosynthesis methyltransferase; Diphthine methyl ester synthase; diphthine synthase; DPH5; DPH5 homolog; HSPC143; NPD015; protein x 0011 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51611 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H2P9 | |
| DPH5 | |
| Recombinant protein corresponding to Human DPH5. Recombinant protein control fragment (Product #RP-108077). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction