missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531963
This item is not returnable.
View return policy
Description
Recommended positive controls: IMR32. Predicted reactivity: Mouse (99%), Rat (100%), Xenopus laevis (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the D4 protein family. The encoded protein is a transcription regulator that binds acetylated histones and is a component of the BAF chromatin remodeling complex. Alternate splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Jul 2013].
Specifications
| DPF3 | |
| Polyclonal | |
| Unconjugated | |
| DPF3 | |
| 2810403B03Rik; 6530402L11Rik; BAF45C; BRG1-associated factor 45C; C78788; CERD4; cer-d4; cerebellum D4; D4 zinc and double PHD finger family 3; D4, zinc and double PHD fingers, family 3; double PHD fingers 3; Dpf3; Zinc finger protein cer-d4; zinc finger protein DPF3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 70127, 8110 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P58269, Q92784 | |
| DPF3 | |
| Recombinant protein encompassing a sequence within the center region of human CERD4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction