missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPF2 Monoclonal Antibody (226CT14.1.1.2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537508
This item is not returnable.
View return policy
Description
DPF2 Monoclonal Antibody for Western Blot
The protein encoded by this gene is a member of the d4 domain family, characterized by a zinc finger-like structural motif. This protein functions as a transcription factor which is necessary for the apoptotic response following deprivation of survival factors. It likely serves a regulatory role in rapid hematopoietic cell growth and turnover. This gene is considered a candidate gene for multiple endocrine neoplasia type I, an inherited cancer syndrome involving multiple parathyroid, enteropancreatic, and pituitary tumors.
Specifications
| DPF2 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.09% sodium azide; pH 7.4 | |
| Q92785 | |
| Dpf2 | |
| DPF2 recombinant protein. | |
| RUO | |
| 100767725, 5977 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 226CT14.1.1.2 | |
| Unconjugated | |
| Dpf2 | |
| 2210010M07Rik; apoptosis response zinc finger protein; BAF45D; BRG1-associated factor 45D; D4, zinc and double PHD fingers family 2; double PHD fingers 2; Dpf2; neuro-d4/requiem; protein requiem; Req; requiem; requiem, apoptosis response zinc finger; UBID4; ubi-d4; Zinc finger protein ubi-d4 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Hamster | |
| Antibody | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction