missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPF2 Antibody (SR1748), Novus Biologicals™
Rabbit Monoclonal Antibody
£282.00 - £367.00
Specifications
| Antigen | DPF2 |
|---|---|
| Clone | SR1748 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604594
|
Novus Biologicals
NBP3-22498-100ul |
100 μg |
£367.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606084
|
Novus Biologicals
NBP3-22498-50ul |
50 μg |
£282.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DPF2 Monoclonal antibody specifically detects DPF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
| DPF2 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Apoptosis response zinc finger protein, D4, zinc and double PHD fingers family 2BAF45D, MGC10180, Protein requiem, REQBRG1-associated factor 45D, requiem, apoptosis response zinc finger, ubi-d4, UBID4requiem, apoptosis response zinc finger gene, zinc finger protein ubi-d4 | |
| A synthesized peptide derived from human DPF2 (Uniprot #: Q92785) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1748 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 5977 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title