missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35810-20ul
This item is not returnable.
View return policy
Description
DPF2 Polyclonal antibody specifically detects DPF2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| DPF2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Apoptosis response zinc finger protein, D4, zinc and double PHD fingers family 2BAF45D, MGC10180, Protein requiem, REQBRG1-associated factor 45D, requiem, apoptosis response zinc finger, ubi-d4, UBID4requiem, apoptosis response zinc finger gene, zinc finger protein ubi-d4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 90-200 of human DPF2 (NP_006259.1).,, Sequence:, DPRLSFPSIKPDTDQTLKKEGLISQDGSSLEALLRTDPLEKRGAPDPRVDDDSLGEFPVTNSRARKRILEPDDFLDDLDDEDYEEDTPKRRGKGKSKGKGVGSARKKLDAS | |
| 20 μL | |
| Apoptosis | |
| 5977 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction