missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97764-100ul
This item is not returnable.
View return policy
Description
DPF2 Polyclonal antibody specifically detects DPF2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| DPF2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 0.5-2 μL/mg of lysate | |
| Apoptosis response zinc finger protein, D4, zinc and double PHD fingers family 2BAF45D, MGC10180, Protein requiem, REQBRG1-associated factor 45D, requiem, apoptosis response zinc finger, ubi-d4, UBID4requiem, apoptosis response zinc finger gene, zinc finger protein ubi-d4 | |
| Produced in rabbits immunized with E. coli-derived Human DPF2 fragment. | |
| 100 μg | |
| Apoptosis | |
| 5977 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion