missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540878
This item is not returnable.
View return policy
Description
Peptide sequence: KELAWVPEAQ RKHTAKKAPD GTVIPNGYCD FCLGGSKKTG CPEDLISCAD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
May have an important role in developing neurons by participating in regulation of cell survival, possibly as a neurospecific transcription factor. Belongs to the neuron-specific chromatin remodeling complex (nBAF complex). During neural development a switch from a stem/progenitor to a post-mitotic chromatin remodeling mechanism occurs as neurons exit the cell cycle and become committed to their adult state. The transition from proliferating neural stem/progenitor cells to post-mitotic neurons requires a switch in subunit composition of the npBAF and nBAF complexes. As neural progenitors exit mitosis and differentiate into neurons, npBAF complexes which contain ACTL6A/BAF53A and PHF10/BAF45A, are exchanged for homologous alternative ACTL6B/BAF53B and DPF1/BAF45B or DPF3/BAF45C subunits in neuron-specific complexes (nBAF). The npBAF complex is essential for the self-renewal/proliferative capacity of the multipotent neural stem cells. The nBAF complex along with CREST plays a role regulating the activity of genes essential for dendrite growth.
Specifications
| DPF1 | |
| Polyclonal | |
| Unconjugated | |
| DPF1 | |
| Baf45b; BRG1-associated factor 45B; D4, zinc and double PHD fingers family 1; double PHD fingers 1; DPF1; NEU BAF45b; NEUD4; neuro-d4; neuro-d4 homolog; neuronal d4 domain family member; zinc finger protein neuro-d4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 50545, 8193 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P56163, Q92782 | |
| DPF1 | |
| Synthetic peptide directed towards the middle region of human DPF1. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction