missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DPEP3 Recombinant Mouse Monoclonal Antibody (5H3)
GREENER_CHOICE

Mouse Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA560729

Product Code. 30535041

  • £382.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DPEP3 Recombinant Monoclonal Antibody for ELISA

This gene encodes a membrane-bound glycoprotein from the family of dipeptidases involved in hydrolytic metabolism of various dipeptides, including penem and carbapenem beta-lactam antibiotics. This gene is located on chromosome 16 in a cluster with another member of this family. Alternatively spliced transcript variants that encode different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DPEP3
Recombinant Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q9H4B8
DPEP3
Recombinant Human DPEP3 protein.
100 μL
Primary
Human, Monkey
Antibody
IgG1
ELISA
5H3
Unconjugated
DPEP3
1700018F16Rik; AI507039; Dipeptidase 3; DPEP3; Mbd3; MBD-3; Membrane-bound dipeptidase 3; metallopeptidase (family M19); MLZ-136; Protein expressed in male leptotene and zygotene spermatocytes 136; putative membrane-bound dipeptidase 3; testis secretory sperm-binding protein Li 211a; UNQ834/PRO1772
Mouse
Affinity chromatography
RUO
64180
-20°C or -80°C if preferred
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.