missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPEP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572282
This item is not returnable.
View return policy
Description
DPEP2 Polyclonal Antibody for Western Blot
Probable metalloprotease which hydrolyzes leukotriene D4 (LTD4) into leukotriene E4 (LTE4).
Specifications
| DPEP2 | |
| Polyclonal | |
| Unconjugated | |
| DPEP2 | |
| dipeptidase 2; DPEP2; F630103D06Rik; MBD2; MBD-2; Membrane-bound dipeptidase 2; putative membrane-bound dipeptidase 2; UNQ284/PRO323 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 319446, 64174 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8C255, Q9H4A9 | |
| DPEP2 | |
| KLH conjugated synthetic peptide between 220-254 amino acids from the Central region of human DPEP2. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction