missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DPEP2 Polyclonal antibody specifically detects DPEP2 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | DPEP2 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:5000-1:10000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | dipeptidase 2, EC 3.4.13.19, MBD2 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human DPEP2 (Accession#: AAH24021.1; Met1-Ser376) |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?